8-Quinolinol sulfate monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Q0017-25G Q0017-500G
TCQ0017-025GEA 77.74 CAD
TCQ0017-025G TCQ0017-500G
8-Quinolinol sulfate monohydrate ≥98.0% (by titrimetric analysis)
8-Quinolinol sulfate monohydrate

CAS Number: 134-31-6
MDL Number: MFCD00013095
Molecular Formula: C9H7NO
Molecular Weight: 388.39
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 178


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: Chinosol, 8-Hydroxyquinoline Sulfate, Oxine Sulfate, 8-Quinolinone Sulfate

Formula: C₁₈H₁₆N₂O₆S
Melting Pt: 178 °C
MDL Number: MFCD00013095
CAS Number: 134-31-6
EINECS: 205-137-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3ec064105a34ae487634c71a186eb01c0a30747&vid=0c67b48f13a0d509d20a9acd3b8e515ff9bf64be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR