N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide ≥98.0%
Supplier: TCI America
Synonyms:
2-Amino-4,6-dichloro-5-formamidopyrimidine, N-(2-Amino-4,6-dichloro-5-pyrimidinyl)formamide
A2146-25G
A2146-5G
TCA2146-25GEA
185.18
CAD
TCA2146-25G
TCA2146-5G
N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide ≥98.0%
N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide
CAS Number: 171887-03-9
MDL Number: MFCD04112936
Molecular Formula: C5H4Cl2N4O
Molecular Weight: 207.01
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Other Product Names / Synonyms: N-(2-Amino-4,6-dichloro-5-pyrimidinyl)formamide
Formula:
C₅H₄Cl₂N₄O |
MDL Number:
MFCD04112936 CAS Number: 171887-03-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...