Quinaldic acid sodium salt ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Q0004-1G Q0004-25G
TCQ0004-001GEA 104.12 CAD
TCQ0004-001G TCQ0004-025G
Quinaldic acid sodium salt ≥98.0% (by HPLC, titration analysis)
Quinaldic acid sodium salt

CAS Number: 16907-79-2
MDL Number: MFCD00070526
Molecular Formula: C10H7NO2
Molecular Weight: 195.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Sodium Quinaldinate, Sodium 2-Quinolinecarboxylate, 2-Quinolinecarboxylic Acid Sodium Salt

Formula: C₁₀H₇NO₂
MDL Number: MFCD00070526
CAS Number: 16907-79-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6140e0b2040945008e23c0899855081456c8e7f&vid=f6140e0b2040945008e23c0899855081456c8e7f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR