Phenylboronic acid pinacol ester ≥98.0%

Supplier: TCI America

P1855-5G P1855-25G
TCP1855-5GEA 81.35 CAD
TCP1855-5G TCP1855-25G
Phenylboronic acid pinacol ester ≥98.0%

CAS Number: 24388-23-6
MDL Number: MFCD00966985
Molecular Formula: C12H17BO2
Molecular Weight: 204.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 130
Freezing point (°C): 28


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, Phenylboronic Acid Pinacol Ester

Formula: C₁₂H₁₇BO₂
Boiling Pt: 130 °C
MDL Number: MFCD00966985
CAS Number: 24388-23-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebc6b4d197cf39904c514e637f5f5584355f8aaa&vid=ebc6b4d197cf39904c514e637f5f5584355f8aaa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR