2-Amino-6-chloropyrazine ≥98.0%
Supplier: TCI America
A2145-1G
A2145-5G
TCA2145-1GEA
39.5
CAD
TCA2145-1G
TCA2145-5G
2-Amino-6-chloropyrazine ≥98.0%
2-Amino-6-chloropyrazine
CAS Number: 33332-28-4
MDL Number: MFCD00055024
Molecular Formula: C4H4ClN3
Molecular Weight: 129.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 153
Formula:
C₄H₄ClN₃ Melting Pt: 153 °C |
MDL Number:
MFCD00055024 CAS Number: 33332-28-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...