Phenoxydiphenylphosphine ≥98.0%

Supplier: TCI America

P1843-5G P1843-25G
TCP1843-5GEA 219.62 CAD
TCP1843-5G TCP1843-25G
Phenoxydiphenylphosphine ≥98.0%
Phenoxydiphenylphosphine

CAS Number: 13360-92-4
Molecular Formula: C18H15OP
Molecular Weight: 278.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 170
Specific Gravity (20/20): 1.14
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Mitsunobu Reaction
Other Product Names / Synonyms: Phenyl Diphenylphosphinite

Formula: C₁₈H₁₅OP
Boiling Pt: 170 °C
Density: 1.14 (20 °C)
MDL Number: MFCD00046055
CAS Number: 13360-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=001ddb3d9940e619381427bfa6f801c446400905&vid=001ddb3d9940e619381427bfa6f801c446400905&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR