Phenyl-2,3,4-tri-O-benzyl-1-thio-β-L-fucopyranoside ≥98.0%

Supplier: TCI America

P1842-5G P1842-1G
TCP1842-5GEA 1273.48 CAD
TCP1842-5G TCP1842-1G
Phenyl-2,3,4-tri-O-benzyl-1-thio-β-L-fucopyranoside ≥98.0%
Phenyl-2,3,4-tri-O-benzyl-1-thio-beta-L-fucopyranoside

CAS Number: 167612-35-3
Molecular Formula: C33H34O4S
Molecular Weight: 526.69
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 108
Storage Temperature: 0-10°C

Formula: C₃₃H₃₄O₄S
MW: 526.69 g/mol
Melting Pt: 108 °C
CAS Number: 167612-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ddf1d04c342c0266451f686d901838558723a7a&vid=3ddf1d04c342c0266451f686d901838558723a7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR