3-Phenylimidazo[1,5-a]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P1838-5G P1838-25G
TCP1838-5GEA 361.82 CAD
TCP1838-5G TCP1838-25G
3-Phenylimidazo[1,5-a]pyridine ≥98.0% (by GC, titration analysis)
3-Phenylimidazo[1,5-a]pyridine

CAS Number: 35854-46-7
Molecular Formula: C13H10N2
Molecular Weight: 194.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Slightly Pale Yellow
Boiling point (°C): 140
Melting point (°C): 110

Formula: C₁₃H₁₀N₂
Boiling Pt: 140 °C
Melting Pt: 110 °C
CAS Number: 35854-46-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2bbaff01712ba386931b936f01987dce4d45445&vid=f2bbaff01712ba386931b936f01987dce4d45445&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR