2,2,3,3,3-Pentafluoropropyl-p-toluenesulfonate ≥98.0%
Supplier: TCI America
P1834-25G
P1834-5G
TCP1834-25GEA
848.46
CAD
TCP1834-25G
TCP1834-5G
2,2,3,3,3-Pentafluoropropyl-p-toluenesulfonate ≥98.0%
2,2,3,3,3-Pentafluoropropyl-p-toluenesulfonate
CAS Number: 565-42-4
MDL Number: MFCD02093346
Molecular Formula: C10H9F5O3S
Molecular Weight: 304.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 75
Melting point (°C): 54
Flash Point (°C): 109
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Other Fluorinated Group Introducing Agents
Other Product Names / Synonyms: p-Toluenesulfonic Acid 2,2,3,3,3-Pentafluoropropyl Ester, 2,2,3,3,3-Pentafluoropropyl Tosylate
Formula:
C₁₀H₉F₅O₃S Boiling Pt: 75 °C Melting Pt: 54 °C Flash Pt: 109 °C |
MDL Number:
MFCD02093346 CAS Number: 565-42-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...