(R)-Piperidine-2-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: H-D-Homopro-OH, D(+)-Pipecolinic acid, (R)-(+)-2-Piperidinecarboxylic Acid, H-D-HoPro-OH, D-Pipecolinic Acid, D-Homoproline

P1830-25G P1830-5G
TCP1830-25GEA 219.78 CAD
TCP1830-25G TCP1830-5G
(R)-Piperidine-2-carboxylic acid ≥98.0%
(R)-Piperidine-2-carboxylic acid

CAS Number: 1723-00-8
MDL Number: MFCD00064346
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 270
Specific rotation [a]20/D: 27 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: D-Homoproline, D-Pipecolinic Acid, (R)-(+)-2-Piperidinecarboxylic Acid

Formula: C₆H₁₁NO₂
Melting Pt: 270 °C
MDL Number: MFCD00064346
CAS Number: 1723-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b2bb50b2e811b290ee4487298a42f51e6a3ae2b&vid=2b2bb50b2e811b290ee4487298a42f51e6a3ae2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR