2-Phenoxyethyl isobutyrate ≥98.0%
Supplier: TCI America
P1822-25G
P1822-500G
TCP1822-25GEA
52.14
CAD
TCP1822-25G
TCP1822-500G
2-Phenoxyethyl isobutyrate ≥98.0%
2-Phenoxyethyl isobutyrate
CAS Number: 103-60-6
MDL Number: MFCD00027363
Molecular Formula: C12H16O3
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 127
Flash Point (°C): 113
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: Isobutyric Acid 2-Phenoxyethyl Ester
Formula:
C₁₂H₁₆O₃ Boiling Pt: 127 °C Density: 1.05 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00027363 CAS Number: 103-60-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...