Potassium(2-methylphenyl)trifluoroborate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (2-Methylphenyl)trifluoroboric acid potassium salt, Potassium trifluoro(o-tolyl)borate

P1818-1G P1818-5G
TCP1818-1GEA 181.7 CAD
TCP1818-1G TCP1818-5G
Potassium(2-methylphenyl)trifluoroborate ≥98.0% (by HPLC)
Potassium (2-methylphenyl)trifluoroborate

CAS Number: 274257-34-0
MDL Number: MFCD03790006
Molecular Formula: C7H7BF3K
Molecular Weight: 198.04
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 226

Formula: C₇H₇BF₃K
MW: 198.04 g/mol
Melting Pt: 226 °C
MDL Number: MFCD03790006
CAS Number: 274257-34-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e933cebe66697dd66a0c892e49cf8bb497bc4f13&vid=e933cebe66697dd66a0c892e49cf8bb497bc4f13&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR