1,1'-(2,6-Pyridinediyl)bis(3-methylimidazolium)dibromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1,1'-(Pyridine-2,6-diyl)bis(3-methyl-1H-imidazol-3-ium) bromide

P1816-1G P1816-5G
TCP1816-1GEA 171.9 CAD
TCP1816-1G TCP1816-5G
1,1'-(2,6-Pyridinediyl)bis(3-methylimidazolium)dibromide ≥98.0% (by HPLC, titration analysis)
1,1'-(2,6-Pyridinediyl)bis(3-methylimidazolium)dibromide

CAS Number: 263874-05-1
Molecular Formula: C13H15Br2N5
Molecular Weight: 401.11
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Very Pale Yellow

Formula: C₁₃H₁₅Br₂N₅
MW: 401.11 g/mol
MDL Number: MFCD11975436
CAS Number: 263874-05-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f91fc51cc9dbf744fe95b51a1a1808fbb86b72a7&vid=f91fc51cc9dbf744fe95b51a1a1808fbb86b72a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR