Potassium(iodomethyl)trifluoroborate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1808-1G P1808-5G
TCP1808-1GEA 101.12 CAD
TCP1808-1G TCP1808-5G
Potassium(iodomethyl)trifluoroborate ≥98.0% (by titrimetric analysis)
Potassium (iodomethyl)trifluoroborate

CAS Number: 888711-47-5
Molecular Formula: CH2BF3IK
Molecular Weight: 247.83
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 242

Formula: CH₂BF₃IK
Melting Pt: 242 °C
MDL Number: MFCD11112187
CAS Number: 888711-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35a14a25dd597021913c1f789d6124df253cfc3d&vid=35a14a25dd597021913c1f789d6124df253cfc3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR