Pyrrolidin-2-ylmethanol ≥95.0% (by GC, titration analysis)

Supplier: TCI America

P1793-5G P1793-25G
TCP1793-5GEA 178.54 CAD
TCP1793-5G TCP1793-25G
Pyrrolidin-2-ylmethanol ≥95.0% (by GC, titration analysis)
Pyrrolidin-2-ylmethanol

CAS Number: 498-63-5
MDL Number: MFCD00601073
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 76
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 2-(Hydroxymethyl)pyrrolidine, 2-Pyrrolidinemethanol

Formula: C5H11NO
Boiling Pt: 76 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00601073
CAS Number: 498-63-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6d8698958612481204b72d06d82ba2ce5a39e0b&vid=e6d8698958612481204b72d06d82ba2ce5a39e0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR