(±)-β-Aminophenethyl alcohol ≥98.0%
Supplier: TCI America
Synonyms:
DL-2-Phenylglycinol, 2-Amino-2-phenylethanol, DL-Phenylglycinol
P1792-5G
P1792-25G
TCP1792-5GEA
123.24
CAD
TCP1792-5G
TCP1792-25G
(±)-β-Aminophenethyl alcohol ≥98.0%
(±)-β-Aminophenethyl alcohol
CAS Number: 7568-92-5
MDL Number: MFCD00130145
Molecular Formula: C8H11NO
Molecular Weight: 137.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 106
Melting point (°C): 51
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Amino-2-phenylethanol
Formula:
C₈H₁₁NO Boiling Pt: 106 °C Melting Pt: 51 °C |
MDL Number:
MFCD00130145 CAS Number: 7568-92-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...