(S)-5-(Pyrrolidin-2-yl)-1H-tetrazole ≥98.0% (by titrimetric analysis)
Supplier: TCI America
P1784-100MG
P1784-500MG
TCP1784-100MGEA
118.5
CAD
TCP1784-100MG
TCP1784-500MG
(S)-5-(Pyrrolidin-2-yl)-1H-tetrazole ≥98.0% (by titrimetric analysis)
(S)-5-(Pyrrolidin-2-yl)-1H-tetrazole
CAS Number: 33878-70-5
Molecular Formula: C5H9N5
Molecular Weight: 139.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 271
Specific rotation [a]20/D: -8 deg (C=1, MeOH)
Formula:
C₅H₉N₅ Melting Pt: 271 °C |
MDL Number:
MFCD10000886 CAS Number: 33878-70-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...