Piperitone (mixture of enantiomers) (predominantly (R)-(-)-form) ≥94.0%

Supplier: TCI America

Synonyms: 6-Isopropyl-3-methyl-2-cyclohexen-1-one, p-Menth-1-en-3-one

P1783-25G P1783-100G
TCP1783-25GEA 93.22 CAD
TCP1783-25G TCP1783-100G
Piperitone (mixture of enantiomers) (predominantly (R)-(-)-form) ≥94.0%
Piperitone (mixture of enantiomers)

CAS Number: 89-81-6
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 235
Melting point (°C): -29
Flash Point (°C): 93
Specific Gravity (20/20): 0.94
Specific rotation [a]20/D: -50 deg (neat)


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: p-Menth-1-en-3-one, 6-Isopropyl-3-methyl-2-cyclohexen-1-one

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 235 °C
Melting Pt: –29 °C
Density: 0.94 (20 °C)
Flash Pt: 93 °C
CAS Number: 89-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1030a6ac3c43053387e079e70885813d7a07219e&vid=19514ab2b4abd380b26ce8b73780d1888886e1c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR