Polyethylene glycol monocetyl ether, M.W. n=∼23 for biochemical research
Supplier: TCI America
P1776-25G
P1776-5G
TCP1776-25GEA
112.18
CAD
TCP1776-25G
TCP1776-5G
Polyethylene glycol monocetyl ether, M.W. n=∼23 for biochemical research
Polyethylene glycol monocetyl ether
[for Biochemical Research]
CAS Number: 9004-95-9
MDL Number: MFCD00080892
Form: Crystal
Color: White
Flash Point (°C): 150
Other Product Names / Synonyms: Polyethylene Glycol Monohexadecyl Ether, Polyoxyethylene Monocetyl Ether
Formula:
HO(CH₂CH₂O)nC₁₆H₃₃ Flash Pt: 150 °C |
MDL Number:
MFCD00080892 CAS Number: 9004-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...