3-Pyridinecarboxylic anhydride ≥97.0%

Supplier: TCI America

Synonyms: Nicotinic Anhydride

P1768-1G P1768-5G
TCP1768-1GEA 142.2 CAD
TCP1768-1G TCP1768-5G
3-Pyridinecarboxylic anhydride ≥97.0%
3-Pyridinecarboxylic anhydride

CAS Number: 16837-38-0
Molecular Formula: C12H8N2O3
Molecular Weight: 228.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 200
Melting point (°C): 124


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: Nicotinic Anhydride

Formula: C₁₂H₈N₂O₃
Boiling Pt: 200 °C
Melting Pt: 124 °C
CAS Number: 16837-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1292f3afe00caeae37c4c685b63257e296aa780e&vid=1292f3afe00caeae37c4c685b63257e296aa780e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR