Phytosphingosine ≥98.0%

Supplier: TCI America

Synonyms: (2S,3S,4R)-2-Aminooctadecane-1,3,4-triol, (2S,3S,4R)-2-Amino-1,3,4-octadecanetriol

P1765-1G P1765-5G
TCP1765-1GEA 192.9 CAD
TCP1765-1G TCP1765-5G
Phytosphingosine ≥98.0%
Phytosphingosine

CAS Number: 554-62-1
MDL Number: MFCD00079232
Molecular Formula: C18H39NO3
Molecular Weight: 317.51
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 102
Specific rotation [a]20/D: 9 deg (C=1, Pyridine)


Product Categories: Bioscience, Lipids, Cosmetic Ingredients for Research and Experimental Use
Other Product Names / Synonyms: (2S,3S,4R)-2-Amino-1,3,4-octadecanetriol

Formula: C₁₈H₃₉NO₃
Melting Pt: 102 °C
MDL Number: MFCD00079232
CAS Number: 554-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f20d293aa6e177d6e39326787eb2d817935a57af&vid=f20d293aa6e177d6e39326787eb2d817935a57af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR