5-Pyrimidineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Pyrimidine-5-boronic acid, 5-Pyrimidylboronic acid, Pyrimidin-5-ylboronic acid, 5-Pyrimidinylboronic acid

P1759-100MG P1759-1G
TCP1759-100MGEA 52.14 CAD
TCP1759-100MG TCP1759-1G
5-Pyrimidineboronic acid (contains varying amounts of Anhydride)
5-Pyrimidineboronic acid

CAS Number: 109299-78-7
MDL Number: MFCD03002366
Molecular Formula: C4H5BN2O2
Molecular Weight: 123.91
Form: Crystal
Color: White
Melting point (°C): 114
Storage Temperature: 0-10°C

Formula: C₄H₅BN₂O₂
MW: 123.91 g/mol
Melting Pt: 114 °C
MDL Number: MFCD03002366
CAS Number: 109299-78-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea0c98c640d79cfc993e7881d6e578da555d536a&vid=ea0c98c640d79cfc993e7881d6e578da555d536a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR