5-Phenyl-3-(trifluoromethyl)-1H-pyrazole ≥98.0%

Supplier: TCI America

Synonyms: 5-Phenyl-3-(trifluoromethyl)pyrazole

P1756-1G P1756-5G
TCP1756-1GEA 61.62 CAD
TCP1756-1G TCP1756-5G
5-Phenyl-3-(trifluoromethyl)-1H-pyrazole ≥98.0%
5-Phenyl-3-(trifluoromethyl)-1H-pyrazole

CAS Number: 4027-54-7
MDL Number: MFCD00156064
Molecular Formula: C10H7F3N2
Molecular Weight: 212.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 124

Formula: C₁₀H₇F₃N₂
Melting Pt: 124 °C
MDL Number: MFCD00156064
CAS Number: 4027-54-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56b4d8d681efa44a77bce5e6f06971ca761cc1a9&vid=56b4d8d681efa44a77bce5e6f06971ca761cc1a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR