1-Amino-4-bromoanthraquinone-2-sulfonic acid sodium salt ≥98.0%

Supplier: TCI America

Synonyms: 1-Amino-4-bromo-9,10-dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt, 1-Amino-4-bromoanthraquinone-2-sulphonic Acid Sodium Salt, Sodium 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate, Bromaminic acid Sodium Salt, Bromaminic acid sodium salt, 1-Amino-4-bromoanthraquinone-2-sulfonic Acid Sodium Salt, Sodium 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate

A0279-25G A0279-500G
TCA0279-025GEA 86.9 CAD
TCA0279-025G TCA0279-500G
1-Amino-4-bromoanthraquinone-2-sulfonic acid sodium salt ≥98.0%
1-Amino-4-bromoanthraquinone-2-sulfonic acid sodium salt

CAS Number: 6258-06-6
MDL Number: MFCD00019160
Molecular Formula: C14H8BrNO5S
Molecular Weight: 404.17
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: Bromaminic Acid Sodium Salt

Formula: C₁₄H₈BrNO₅S
MDL Number: MFCD00019160
CAS Number: 6258-06-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9590d13efcf9ecd1af17bb71b57dc371f0855b97&vid=9590d13efcf9ecd1af17bb71b57dc371f0855b97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR