2-Phenyl-2-(2-pyridyl)acetamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Phenyl-2-(pyridin-2-yl)acetamide

P1741-5G P1741-25G
TCP1741-5GEA 102.54 CAD
TCP1741-5G TCP1741-25G
2-Phenyl-2-(2-pyridyl)acetamide ≥98.0% (by HPLC, titration analysis)
2-Phenyl-2-(2-pyridyl)acetamide

CAS Number: 7251-52-7
MDL Number: MFCD00138126
Molecular Formula: C13H12N2O
Molecular Weight: 212.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 136

Formula: C₁₃H₁₂N₂O
MW: 212.25 g/mol
Melting Pt: 136 °C
MDL Number: MFCD00138126
CAS Number: 7251-52-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4a2d10dc76248653790896c44d5e4df08121d65&vid=a4a2d10dc76248653790896c44d5e4df08121d65&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR