D-Proline benzyl ester hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: H-D-Pro-OBzl.HCl

P1726-1G P1726-5G P1726-25G
TCP1726-1GEA 85.32 CAD
TCP1726-1G TCP1726-5G TCP1726-25G
D-Proline benzyl ester hydrochloride ≥98.0%
D-Proline benzyl ester hydrochloride

CAS Number: 53843-90-6
MDL Number: MFCD00153458
Molecular Formula: C12H15NO2
Molecular Weight: 241.72
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Specific rotation [a]20/D: 44 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Pro-OBzl.HCl

Formula: C₁₂H₁₅NO₂
MW: 241.72 g/mol
MDL Number: MFCD00153458
CAS Number: 53843-90-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=549f7ff8e8149475e006d931f682c3ce185c192b&vid=549f7ff8e8149475e006d931f682c3ce185c192b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR