D-Phenylalanine benzyl ester-p-toluenesulfonate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: H-D-Phe-OBzl.Tos, H-D-Phe-OBzl.TosOH, D-Phenylalanine benzyl ester tosylate

P1722-25G P1722-5G
TCP1722-25GEA 109.02 CAD
TCP1722-25G TCP1722-5G
D-Phenylalanine benzyl ester-p-toluenesulfonate ≥98.0% (by HPLC, titration analysis)
D-Phenylalanine benzyl ester-p-toluenesulfonate

CAS Number: 28607-46-7
MDL Number: MFCD00066129
Molecular Formula: C16H17NO2
Molecular Weight: 427.52
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 169
Specific rotation [a]20/D: 6 deg (C=2, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Phe-OBzl.Tos-OH

Formula: C₂₃H₂₅NO₅S
Melting Pt: 169 °C
MDL Number: MFCD00066129
CAS Number: 28607-46-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a979e79446c94f9f1ec738bbf9c12b33292084b5&vid=d7f1575525b0638f6b728b0262071d9a6079da1d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR