Potassium(3-pyridyl)trifluoroborate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Potassium trifluoro(pyridin-3-yl)borate

P1717-1G P1717-5G
TCP1717-1GEA 109.02 CAD
TCP1717-1G TCP1717-5G
Potassium(3-pyridyl)trifluoroborate ≥98.0% (by total nitrogen basis)
Potassium (3-pyridyl)trifluoroborate

CAS Number: 561328-69-6
Molecular Formula: C5H5BF3N
Molecular Weight: 185.00
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White

Formula: C₅H₄BF₃KN
MW: 185.0000 g/mol
MDL Number: MFCD09038545
CAS Number: 561328-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff439a2bb15f0edb4c82b297837e1b8c7f4975da&vid=ff439a2bb15f0edb4c82b297837e1b8c7f4975da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR