2-Phenylethanethiol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
P1715-25G
P1715-5G
TCP1715-25GEA
281.24
CAD
TCP1715-25G
TCP1715-5G
2-Phenylethanethiol ≥97.0% (by GC, titration analysis)
2-Phenylethanethiol
CAS Number: 4410-99-5
MDL Number: MFCD00004891
Molecular Formula: C8H10S
Molecular Weight: 138.23
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 218
Flash Point (°C): 90
Specific Gravity (20/20): 1.03
Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: Phenethyl Mercaptan, 2-Phenylethyl Mercaptan
Formula:
C₈H₁₀S Boiling Pt: 218 °C Density: 1.03 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00004891 CAS Number: 4410-99-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...