Pentadecyl acetate ≥95.0%

Supplier: TCI America

P1700-25G P1700-5G
TCP1700-25GEA 314.1 CAD
TCP1700-25G TCP1700-5G
Pentadecyl acetate ≥95.0%
Pentadecyl acetate

CAS Number: 629-58-3
MDL Number: MFCD00056204
Molecular Formula: C17H34O2
Molecular Weight: 270.46
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 230
Melting point (°C): 15
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Acetic Acid Pentadecyl Ester

Formula: C₁₇H₃₄O₂
Boiling Pt: 230 °C
Melting Pt: 15 °C
Density: 0.86 (20 °C)
MDL Number: MFCD00056204
CAS Number: 629-58-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e87a1b383786194416bab9a601683669558f5b0&vid=7e87a1b383786194416bab9a601683669558f5b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR