4,4-Pentamethylene-2-pyrrolidone ≥98.0%
Supplier: TCI America
P1695-25G
P1695-5G
TCP1695-25GEA
188.49
CAD
TCP1695-25G
TCP1695-5G
4,4-Pentamethylene-2-pyrrolidone ≥98.0%
4,4-Pentamethylene-2-pyrrolidone
CAS Number: 64744-50-9
MDL Number: MFCD00177938
Molecular Formula: C9H15NO
Molecular Weight: 153.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 181
Melting point (°C): 91
Other Product Names / Synonyms: 3-Azaspiro[4.5]decan-2-one
Formula:
C₉H₁₅NO Boiling Pt: 181 °C Melting Pt: 91 °C |
MDL Number:
MFCD00177938 CAS Number: 64744-50-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...