Potassium trifluoro(trifluoromethyl)borate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

P1692-25G P1692-5G P1692-1G
TCP1692-25GEA 1300.34 CAD
TCP1692-25G TCP1692-5G TCP1692-1G
Potassium trifluoro(trifluoromethyl)borate ≥98.0% (by titrimetric analysis)
Potassium trifluoro(trifluoromethyl)borate

CAS Number: 42298-15-7
Molecular Formula: CHBF6
Molecular Weight: 175.91
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: CHBF₆
MDL Number: MFCD09038544
CAS Number: 42298-15-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e244b010f08f5af54e7a0981d5df44c8fa6673e5&vid=e244b010f08f5af54e7a0981d5df44c8fa6673e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR