1-Pyrenecarboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

P1687-1G P1687-5G
TCP1687-1GEA 120.71 CAD
TCP1687-1G TCP1687-5G
1-Pyrenecarboxylic acid ≥97.0% (by GC, titration analysis)
1-Pyrenecarboxylic acid

CAS Number: 19694-02-1
MDL Number: MFCD00192424
Molecular Formula: C17H10O2
Molecular Weight: 246.27
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 274
Lambda max.: 380 nm (MeOH)

Formula: C₁₇H₁₀O₂
Melting Pt: 274 °C
MDL Number: MFCD00192424
CAS Number: 19694-02-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0267c746a89d973b630e77cb426647b95d3d90b0&vid=0267c746a89d973b630e77cb426647b95d3d90b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR