Potassium (4-pyridyl)trifluoroborate ≥98.0%

Supplier: TCI America

Synonyms: Potassium 4-Pyridyltrifluoroborate

P1684-1G P1684-5G
TCP1684-1GEA 94.8 CAD
TCP1684-1G TCP1684-5G
Potassium (4-pyridyl)trifluoroborate ≥98.0%
Potassium (4-pyridyl)trifluoroborate

CAS Number: 1111732-87-6
Molecular Formula: C5H4BF3KN
Molecular Weight: 185.00
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₅H₄BF₃KN
MW: 184.9965 g/mol
MDL Number: MFCD09038543
CAS Number: 1111732-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bffea16bbdd07358f693ad86404140e2e876561a&vid=bffea16bbdd07358f693ad86404140e2e876561a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR