1-Phenyl-1-hexanol ≥98.0%

Supplier: TCI America

P1683-25G P1683-5G
TCP1683-25GEA 301.78 CAD
TCP1683-25G TCP1683-5G
1-Phenyl-1-hexanol ≥98.0%
1-Phenyl-1-hexanol

CAS Number: 4471-05-0
MDL Number: MFCD02258728
Molecular Formula: C12H18O
Molecular Weight: 178.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 172
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: alpha-Pentylbenzyl Alcohol

Formula: C₁₂H₁₈O
Boiling Pt: 172 °C
Density: 0.95 (20 °C)
MDL Number: MFCD02258728
CAS Number: 4471-05-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d58f57d97a9bc37cd6f0f678cbd8e2c4400cc865&vid=d58f57d97a9bc37cd6f0f678cbd8e2c4400cc865&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR