1-Phenyl-1-butanol ≥98.0%
Supplier: TCI America
P1682-5G
P1682-25G
TCP1682-5GEA
113.76
CAD
TCP1682-5G
TCP1682-25G
1-Phenyl-1-butanol ≥98.0%
1-Phenyl-1-butanol
CAS Number: 614-14-2
MDL Number: MFCD00066173
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 170
Melting point (°C): 16
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: alpha-Propylbenzyl Alcohol
Formula:
C₁₀H₁₄O Boiling Pt: 170 °C Melting Pt: 16 °C Density: 0.98 (20 °C) |
MDL Number:
MFCD00066173 CAS Number: 614-14-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...