Phytyl acetate (cis and trans mixture) ≥88.0%

Supplier: TCI America

Synonyms: 3,7,11,15-Tetramethyl-2-hexadecenyl Acetate, Acetic Acid 3,7,11,15-Tetramethyl-2-hexadecenyl Ester

P1675-25G P1675-5G
TCP1675-25GEA 613.04 CAD
TCP1675-25G TCP1675-5G
Phytyl acetate (cis and trans mixture) ≥88.0%
Phytyl acetate (cis and trans mixture)

CAS Number: 10236-16-5
Molecular Formula: C22H42O2
Molecular Weight: 338.58
Purity/Analysis Method: >88.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 184
Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Diterpenes
Other Product Names / Synonyms: Acetic Acid 3,7,11,15-Tetramethyl-2-hexadecenyl Ester, 3,7,11,15-Tetramethyl-2-hexadecenyl Acetate

Formula: C₂₂H₄₂O₂
MW: 338.58 g/mol
Density: 0.87 (20 °C)
Flash Pt: 184 °C
CAS Number: 10236-16-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42368fac158e559d06cd7110f8bc5b1f3d78fb37&vid=42368fac158e559d06cd7110f8bc5b1f3d78fb37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 88.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR