4-(2-Pyridyl)benzaldehyde ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Pyridin-2-yl-benzaldehyde, 4-(Pyridin-2-yl)benzaldehyde

P1671-1G P1671-5G
TCP1671-1GEA 25.72 CAD
TCP1671-1G TCP1671-5G
4-(2-Pyridyl)benzaldehyde ≥95.0% (by GC, titration analysis)
4-(2-Pyridyl)benzaldehyde

CAS Number: 127406-56-8
MDL Number: MFCD01863537
Molecular Formula: C12H9NO
Molecular Weight: 183.21
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 50
Flash Point (°C): 113


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 2-(4-Formylphenyl)pyridine

Formula: C₁₂H₉NO
MW: 183.21 g/mol
Melting Pt: 50 °C
Flash Pt: 113 °C
MDL Number: MFCD01863537
CAS Number: 127406-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc054862ec7a72afae2657d5ab113d80447317b2&vid=cc054862ec7a72afae2657d5ab113d80447317b2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR