4-Propylcyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

P1656-5G P1656-25G
TCP1656-5GEA 165.9 CAD
TCP1656-5G TCP1656-25G
4-Propylcyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)
4-Propylcyclohexanecarboxylic acid (cis and trans mixture)

CAS Number: 70928-91-5
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless

Formula: C₁₀H₁₈O₂
CAS Number: 70928-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ddb27b4ff9fd06a6d922f8485140f1c1cb59c7&vid=58ddb27b4ff9fd06a6d922f8485140f1c1cb59c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR