4-(4-Pyridyl)morpholine ≥98.0%

Supplier: TCI America

Synonyms: 4-Morpholinopyridine, 4-(Pyridin-4-yl)morpholine

P1646-5G P1646-25G
TCP1646-5GEA 252.8 CAD
TCP1646-5G TCP1646-25G
4-(4-Pyridyl)morpholine ≥98.0%
4-(4-Pyridyl)morpholine

CAS Number: 2767-91-1
Molecular Formula: C9H12N2O
Molecular Weight: 164.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 107


Other Product Names / Synonyms: 4-Morpholinopyridine

Formula: C₉H₁₂N₂O
MW: 164.21 g/mol
Melting Pt: 107 °C
MDL Number: MFCD00704054
CAS Number: 2767-91-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9388b1a1d35bd07d6643958ad75488a86433e21&vid=e9388b1a1d35bd07d6643958ad75488a86433e21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR