cis-3-Aminocyclohexanecarboxylic acid ≥96.0% (by GC, titration analysis)

Supplier: TCI America

A2127-25G A2127-5G
TCA2127-25GEA 1287.7 CAD
TCA2127-25G TCA2127-5G
cis-3-Aminocyclohexanecarboxylic acid ≥96.0% (by GC, titration analysis)
cis-3-Aminocyclohexanecarboxylic acid

CAS Number: 16636-51-4
MDL Number: MFCD09753647
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 278

Formula: C₇H₁₃NO₂
Melting Pt: 278 °C
MDL Number: MFCD09753647
CAS Number: 16636-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46642bbba8798e7b438a6b06ff79913206b81d55&vid=46642bbba8798e7b438a6b06ff79913206b81d55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR