5-Propyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione ≥90.0%

Supplier: TCI America

P1635-1G P1635-250MG
TCP1635-1GEA 679.4 CAD
TCP1635-1G TCP1635-250MG
5-Propyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione ≥90.0%
5-Propyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione

CAS Number: 202126-48-5
Molecular Formula: C7H8S5
Molecular Weight: 252.44
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Color: Yellow Red
Melting point (°C): 51

Formula: C₇H₈S₅
MW: 252.44 g/mol
Melting Pt: 51 °C
CAS Number: 202126-48-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0be6005bd1add931d21d319e3d3be3e437ce282&vid=b0be6005bd1add931d21d319e3d3be3e437ce282&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR