Pyridinium trifluoromethanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Pyridinium Triflate

P1627-5G P1627-25G
TCP1627-5GEA 82.16 CAD
TCP1627-5G TCP1627-25G
Pyridinium trifluoromethanesulfonate ≥98.0%
Pyridinium trifluoromethanesulfonate

CAS Number: 52193-54-1
Molecular Formula: C6H6F3NO3S
Molecular Weight: 229.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 224


Other Product Names / Synonyms: Pyridinium Triflate

Formula: C₆H₆F₃NO₃S
MW: 229.17 g/mol
Melting Pt: 224 °C
MDL Number: MFCD00274656
CAS Number: 52193-54-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=361fd8fe38b0957427ea8e67b53e5f96f9f98205&vid=361fd8fe38b0957427ea8e67b53e5f96f9f98205&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR