1-Pyreneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

P1625-1G P1625-5G P1625-25G
TCP1625-1GEA 61.62 CAD
TCP1625-1G TCP1625-5G TCP1625-25G
1-Pyreneboronic acid (contains varying amounts of Anhydride)
1-Pyreneboronic acid

CAS Number: 164461-18-1
Molecular Formula: C16H11BO2
Molecular Weight: 246.07
Form: Crystal
Color: White
Melting point (°C): 251

Formula: C₁₆H₁₁BO₂
Melting Pt: 251 °C
MDL Number: MFCD04974062
CAS Number: 164461-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105a4ca15c7caad467469e9449ae8ebf596063b7&vid=105a4ca15c7caad467469e9449ae8ebf596063b7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR