Phenyl[2-(trimethylsilyl)phenyl]iodonium trifluoromethanesulfonate ≥97.0%

Supplier: TCI America

Synonyms: Phenyl[2-(trimethylsilyl)phenyl]iodonium Triflate

P1620-1G P1620-5G
TCP1620-1GEA 347.13 CAD
TCP1620-1G TCP1620-5G
Phenyl[2-(trimethylsilyl)phenyl]iodonium trifluoromethanesulfonate ≥97.0%
Phenyl[2-(trimethylsilyl)phenyl]iodonium trifluoromethanesulfonate

CAS Number: 164594-13-2
Molecular Formula: C16H18F3IO3SSi
Molecular Weight: 502.36
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 132


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Hypervalent Iodine Compounds, Benzyne Precursors (Multi-component Reaction), Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: Phenyl[2-(trimethylsilyl)phenyl]iodonium Triflate

Formula: C₁₆H₁₈F₃IO₃SSi
MW: 502.36 g/mol
Melting Pt: 132 °C
CAS Number: 164594-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6369c0a8b59bd64b1a4f16eb30c27336d58beeb5&vid=6369c0a8b59bd64b1a4f16eb30c27336d58beeb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR