1,3-Propanediamine-N,N,N',N'-tetraacetic acid ≥98.0%
Supplier: TCI America
Synonyms:
Trimethylenediamine-N,N,N',N'-tetraacetic Acid, 2,2',2'',2'''-(Propane-1,3-diylbis(azanetriyl))tetraacetic acid
P1605-25G
P1605-500G
TCP1605-25GEA
48.33
CAD
TCP1605-25G
TCP1605-500G
1,3-Propanediamine-N,N,N',N'-tetraacetic acid ≥98.0%
1,3-Propanediamine-N,N,N',N'-tetraacetic acid
CAS Number: 1939-36-2
Molecular Formula: C11H18N2O8
Molecular Weight: 306.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 227
Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: Trimethylenediamine-N,N,N',N'-tetraacetic Acid
Formula:
C₁₁H₁₈N₂O₈ MW: 306.27 g/mol Melting Pt: 227 °C |
MDL Number:
MFCD00210039 CAS Number: 1939-36-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...