4-Phenylthiomorpholine-1,1-dioxide ≥98.0%

Supplier: TCI America

P1603-5G P1603-25G
TCP1603-5GEA 205.4 CAD
TCP1603-5G TCP1603-25G
4-Phenylthiomorpholine-1,1-dioxide ≥98.0%
4-Phenylthiomorpholine-1,1-dioxide

CAS Number: 17688-68-5
Molecular Formula: C10H13NO2S
Molecular Weight: 211.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 120

Formula: C₁₀H₁₃NO₂S
MW: 211.28 g/mol
Melting Pt: 120 °C
CAS Number: 17688-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b859fe6cc6ed229d1d43afcdc274e04acefa6bb&vid=9b859fe6cc6ed229d1d43afcdc274e04acefa6bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR