5-(Aminosulfonyl)-2,3-dimethoxybenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A2126-25G A2126-5G
TCA2126-25GEA 159.58 CAD
TCA2126-25G TCA2126-5G
5-(Aminosulfonyl)-2,3-dimethoxybenzoic acid ≥98.0% (by HPLC, titration analysis)
5-(Aminosulfonyl)-2,3-dimethoxybenzoic acid

CAS Number: 66644-80-2
MDL Number: MFCD01317561
Molecular Formula: C9H11NO6S
Molecular Weight: 261.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 206


Other Product Names / Synonyms: 2,3-Dimethoxy-5-sulfamoylbenzoic Acid

Formula: C₉H₁₁NO₆S
Melting Pt: 206 °C
MDL Number: MFCD01317561
CAS Number: 66644-80-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=407abd7528fc742f224e01d310a111a4505a15e7&vid=407abd7528fc742f224e01d310a111a4505a15e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR