1,2,2,6,6-Pentamethyl-4-piperidyl methacrylate ≥97.0% stabilised

Supplier: TCI America

Synonyms: Methacrylic Acid 1,2,2,6,6-Pentamethyl-4-piperidyl Ester

P1513-25G P1513-500G
TCP1513-25GEA 129.56 CAD
TCP1513-25G TCP1513-500G
1,2,2,6,6-Pentamethyl-4-piperidyl methacrylate ≥97.0% stabilised
1,2,2,6,6-Pentamethyl-4-piperidyl methacrylate

(stabilized with MEHQ)
CAS Number: 68548-08-3
Molecular Formula: C14H25NO2
Molecular Weight: 239.36
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 112
Specific Gravity (20/20): 0.97
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Resin Additives, Monomers
Other Product Names / Synonyms: Methacrylic Acid 1,2,2,6,6-Pentamethyl-4-piperidyl Ester

Formula: C₁₄H₂₅NO₂
MW: 239.36 g/mol
Density: 0.97 (20 °C)
Flash Pt: 112 °C
CAS Number: 68548-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3158286b4a237b7da533eeacbf2568f562e995ef&vid=3158286b4a237b7da533eeacbf2568f562e995ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR