(S)-(-)-3-Phenylcyclohexanone ≥98.0%

Supplier: TCI America

P1500-100MG P1500-1G
TCP1500-100MGEA 127.35 CAD
TCP1500-100MG TCP1500-1G
(S)-(-)-3-Phenylcyclohexanone ≥98.0%
(S)-(-)-3-Phenylcyclohexanone

CAS Number: 57344-86-2
Molecular Formula: C12H14O
Molecular Weight: 174.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Freezing point (°C): 22
Specific rotation [a]20/D: -20 deg (C=1, CHCl3)
Storage Temperature: 0-10°C

Formula: C₁₂H₁₄O
MDL Number: MFCD08276411
CAS Number: 57344-86-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9e8cc1b5756a0994d48d9ac2566abbe563c3854&vid=a9e8cc1b5756a0994d48d9ac2566abbe563c3854&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR